Learn More
Perkin Elmer US LLC Organics Sample Introduction Kit for Optima 8x00
Catalog No. 502112266
Encompass_Preferred
This kit includes: 3-Slot Quartz Torch, Qty. 2 (N0780132) 1. 2 mm I.D. Alumina Injector, Qty. 2 (N0781007) Baffled Cyclonic Spray Chamber 4 mm, Qty. 1 (N0776090) GemCone Low-Flow Nebulizer, Qty. 1 (N0770358) Cyclonic Spray Chamber Adapter, Qty. 1 (N0770614) Injector Adapter Washer, Qty. 1 (N0776014) Injector Adapter, Qty. 1 (N0776091) Injector Adapter Nut, Qty. 1 (N0776025) O-Ring 6.07 mm I.D., Qty. 1 (09902207) O-Ring 8.74 mm I.D., Qty. 2 (09921028)
Catalog No. 50-211-2266 Supplier Perkin Elmer US LLC Supplier No. N0780608
This kit includes: 3-Slot Quartz Torch, Qty. 2 (N0780132) 1. 2 mm I.D. Alumina Injector, Qty. 2 (N0781007) Baffled Cyclonic Spray Chamber 4 mm, Qty. 1 (N0776090) GemCone Low-Flow Nebulizer, Qty. 1 (N0770358) Cyclonic Spray Chamber Adapter, Qty. 1 (N0770614) Injector Adapter Washer, Qty. 1 (N0776014) Injector Adapter, Qty. 1 (N0776091) Injector Adapter Nut, Qty. 1 (N0776025) O-Ring 6.07 mm I.D., Qty. 1 (09902207) O-Ring 8.74 mm I.D., Qty. 2 (09921028)