Learn More
Perkin Elmer US LLC Organics Sample Introduction Torch Module for Avio 200
Catalog No. 502113122
Encompass_Preferred
This kit includes: GemCone Low-Flow Nebulizer (N0770358) Injector Adapter Kit-Cyclonic with O-Rings (N0790141) Hybrid XLT 3 Slot Torch Avio (N0790249) Alumina Injector 1.2 mm Length Avio (N0791182) Baffled Cyclonic Spray Chamber Avio (N0791352) Torch Body Assembly for Avio 200 (N0790138)
Catalog No. 50-211-3122 Supplier Perkin Elmer US LLC Supplier No. N0790611
This kit includes: GemCone Low-Flow Nebulizer (N0770358) Injector Adapter Kit-Cyclonic with O-Rings (N0790141) Hybrid XLT 3 Slot Torch Avio (N0790249) Alumina Injector 1.2 mm Length Avio (N0791182) Baffled Cyclonic Spray Chamber Avio (N0791352) Torch Body Assembly for Avio 200 (N0790138)