Learn More
Perkin Elmer US LLC Organics Stainless Steel Nebulizer Complete Assembly for PinAAcle
Catalog No. 502104360
Encompass_Preferred
A complete Stainless Steel body with Stainless Steel nebulizer, Stainless Steel needle, and Kalrez o-rings. It is preferred for general applications with acid concentrations less than 5%, providing exceptional performance and durability. This assembly includes: Solvent Resistant O-Ring Sealing Nebulizer to End Cap, Qty. 1 (09200274) Cap, Qty. 1 (N3161057) Venturi, Qty. 1 (03031810) O-Ring for Venturi, Qty. 1 (09902047) Solvent Resistant O-Rings for Nebulizer Body, Qty. 2 (09200276) Nebulizer Body, Qty. 1 (N3160133) Nebulizer Insert, Qty. 1 (03031812) Solvent Resistant O-Ring for Nebulizer Insert, Qty. 1 (09200273) Locking Ring, Qty. 1 (N3161006) Spring, Qty. 1 (03031806) Solvent Resistant O-Ring for Capillary Needle Assembly, Qty. 1 (09200271) Solvent Resistant Capillary Needle Assembly, Qty. 1 (N3160153) PTFE Washer, Qty. 1 (03031802) Fine Adjustment Nut, Qty. 1 (03031460) Sample Capillary, Qty. 1 (09908265) This nebulizer is compatible with the PerkinElmer PinAAcle 500/900F/900H/90
Catalog No. 50-210-4360 Supplier Perkin Elmer US LLC Supplier No. N3160158
A complete Stainless Steel body with Stainless Steel nebulizer, Stainless Steel needle, and Kalrez o-rings. It is preferred for general applications with acid concentrations less than 5%, providing exceptional performance and durability. This assembly includes: Solvent Resistant O-Ring Sealing Nebulizer to End Cap, Qty. 1 (09200274) Cap, Qty. 1 (N3161057) Venturi, Qty. 1 (03031810) O-Ring for Venturi, Qty. 1 (09902047) Solvent Resistant O-Rings for Nebulizer Body, Qty. 2 (09200276) Nebulizer Body, Qty. 1 (N3160133) Nebulizer Insert, Qty. 1 (03031812) Solvent Resistant O-Ring for Nebulizer Insert, Qty. 1 (09200273) Locking Ring, Qty. 1 (N3161006) Spring, Qty. 1 (03031806) Solvent Resistant O-Ring for Capillary Needle Assembly, Qty. 1 (09200271) Solvent Resistant Capillary Needle Assembly, Qty. 1 (N3160153) PTFE Washer, Qty. 1 (03031802) Fine Adjustment Nut, Qty. 1 (03031460) Sample Capillary, Qty. 1 (09908265) This nebulizer is compatible with the PerkinElmer PinAAcle 500/900F/900H/90