Learn More
Enterprise Technology Solutions The ndd Medical EasyOne FlowTube Mouthpiece
SDP
Catalog No. 502407022
Encompass_Preferred
The ndd Medical EasyOne FlowTube Mouthpiece allows one to avoid cross-contamination and reduce cleaning to a minimum. All parts exposed to a patient's breath are single-use and disposable. EasyOne FlowTubes are made from polypropylen (PP), and there is no expiration date or special storage conditions required.The EasyOne FlowTube is compatible with the EasyOne Air Spirometer.
Catalog No. 50-240-7022 Supplier Enterprise Technology Solutions Supplier No. NDD5050200
The ndd Medical EasyOne FlowTube Mouthpiece allows one to avoid cross-contamination and reduce cleaning to a minimum. All parts exposed to a patient's breath are single-use and disposable. EasyOne FlowTubes are made from polypropylen (PP), and there is no expiration date or special storage conditions required.The EasyOne FlowTube is compatible with the EasyOne Air Spirometer.