Learn More
Med Vet International McKesson Tube Organizer, 5-1/2 X 11-1/2 X 16 Inch, 3118, Each
Catalog No. 503046995
Encompass_Preferred
Tiered Tube Organizer McKesson 9 Place Clear 5-1/2 X 11-1/2 X 16 Inch Three rows and nine sections give lab workers and phlebotomists plenty of options when sorting and stowing tubes Can accommodate multiple sizes and will hold up to 210 tubes Designed to wall mount or for use on a benchtop, these clear, acrylic racks are a perfect drawing station accessory 11½ x 5½ x 16 inches
Catalog No. 50-304-6995 Supplier Med Vet International Supplier No. 883766EA
Tiered Tube Organizer McKesson 9 Place Clear 5-1/2 X 11-1/2 X 16 Inch Three rows and nine sections give lab workers and phlebotomists plenty of options when sorting and stowing tubes Can accommodate multiple sizes and will hold up to 210 tubes Designed to wall mount or for use on a benchtop, these clear, acrylic racks are a perfect drawing station accessory 11½ x 5½ x 16 inches