Learn More
MIMETAS US Organoflow S Perfusion System
SDP
Catalog No. NC1878412
Encompass
Optimal control of perfusion flow in OrganoPlates. Pump and tube-free perfusion of your cultures. Optimized time and tilting angle settings, including interval rocking. Suitable for use in standard tissue culture incubators. Maintain flow in up to 16 OrganoPlates.
Catalog No. NC1878412 Supplier MIMETAS US Supplier No. MIOFPRS
Optimal control of perfusion flow in OrganoPlates. Pump and tube-free perfusion of your cultures. Optimized time and tilting angle settings, including interval rocking. Suitable for use in standard tissue culture incubators. Maintain flow in up to 16 OrganoPlates.