Learn More
STEMCELL Technologies Organoid Embedding Sheet (6-Pack), Size: 6 Units
SDP
Catalog No. NC1692080
Encompass
The Organoid Embedding Sheet is designed to facilitate embedding 3-dimensional (3D) aggregates (e.g. embryoid bodies, spheroids, or organoids) into a matrix. The wells on the sheet are designed to fit aggregates up to 5 mm in diameter. The sheet may be grasped at the top, along the label, for easy handling. The Organoid Embedding Sheet can also be a substitute for Parafilm and used to mix small volumes in various protocols. Each sheet contains 18 wells, and the sheet is compatible with STEMCELL media products such as STEMdiff™ Cerebral Organoid Kit (Catalog #08570).
Catalog No. NC1692080 Supplier STEMCELL Technologies Supplier No. 08579
The Organoid Embedding Sheet is designed to facilitate embedding 3-dimensional (3D) aggregates (e.g. embryoid bodies, spheroids, or organoids) into a matrix. The wells on the sheet are designed to fit aggregates up to 5 mm in diameter. The sheet may be grasped at the top, along the label, for easy handling. The Organoid Embedding Sheet can also be a substitute for Parafilm and used to mix small volumes in various protocols. Each sheet contains 18 wells, and the sheet is compatible with STEMCELL media products such as STEMdiff™ Cerebral Organoid Kit (Catalog #08570).