Learn More
ATCC Organoid Growth Kit 1A
Catalog No. 502384635
Encompass
ATCC Growth Kits are packages of recombinant proteins, small molecules, and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in a convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.
Catalog No. 50-238-4635 Supplier ATCC Supplier No. ACS7100
ATCC Growth Kits are packages of recombinant proteins, small molecules, and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in a convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.