Learn More
ATCC Organoid Growth Kit 1B
Catalog No. 502384689
Encompass
ATCC Growth Kits are packages of recombinant proteins, small molecules and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.
Catalog No. 50-238-4689 Supplier ATCC Supplier No. ACS7101
 Restricted Item
Add to Cart
Add to Cart
ATCC Growth Kits are packages of recombinant proteins, small molecules and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.