Learn More
ATCC Organoid Growth Kit 1F
Catalog No. 502384362
Encompass
ATCC Growth Kits are packages of recombinant proteins, small molecules and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.
Catalog No. 50-238-4362 Supplier ATCC Supplier No. ACS7105
Add to Cart
Add to Cart
ATCC Growth Kits are packages of recombinant proteins, small molecules and other supplements designed to make the preparation of complex media formulations for select organoid culture models easy and reliable. Kit components are supplied as powders to maximize stability and in convenient, pre-portioned, ready-to-reconstitute format that does not require aliquoting or storage once prepared.