Learn More
Innovative Research Inc Innovative Grade US Orgin Sheep Whole Blood Defibrinated 100ml
Catalog No. 502034461
Encompass_Preferred
Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color. We offer reliable, consistent products that deliver reliable, consistent results.
Catalog No. 50-203-4461 Supplier Innovative Research Inc Supplier No. IGSHWBDF100ML
Innovative Grade US Origin Sheep Whole Blood Defibrinated from Innovative Research is collected per-order from sheep located in the United States. The animals used for this material are of US Origin, ensuring that only the highest-quality material is used. The whole blood is mechanically defibrinated without anticoagulants or other additives. Our proprietary screening process ensures low fragility and consistent color. We offer reliable, consistent products that deliver reliable, consistent results.