Learn More
MSC Rite-Kem Organic Solvent Degreaser, 4 x 1 gal, Liquid Concentrate, Biodegradable
Catalog No. 502835722
Encompass
This organic solvent degreaser is distilled from all organic plant material, making it 100% free from petroleum solvents. It is a liquid concentrate designed for multiple uses as an all-purpose cleaner and degreaser.
  • All-purpose cleaner and degreaser
  • Liquid concentrate
  • 100% free from petroleum solvents
  • Biodegradable formula
  • Water-based solution
  • Pleasant scent
  • Dilutable
Catalog No. 50-283-5722 Supplier MSC Supplier No. 48175939
This organic solvent degreaser is distilled from all organic plant material, making it 100% free from petroleum solvents. It is a liquid concentrate designed for multiple uses as an all-purpose cleaner and degreaser.
  • All-purpose cleaner and degreaser
  • Liquid concentrate
  • 100% free from petroleum solvents
  • Biodegradable formula
  • Water-based solution
  • Pleasant scent
  • Dilutable