Learn More
MSC Rite-Kem Organic Solvent Degreaser, 55 gal, Plant-Based, Petroleum-Free
Catalog No. 502835721
Encompass
This organic solvent degreaser is a plant-based, 100% petroleum-free, liquid concentrate with a citrus scent. It is a dilutable, water-based, and biodegradable formula designed for various cleaning applications.
  • Liquid form
  • Plastic drum container
  • 55 gallon container size
  • Citrus scent
  • Dilutable concentrate solution
  • Biodegradable, water-based formula
  • 40% VOC content
  • Distilled from all organic plant material
  • 100% free from petroleum solvents
Catalog No. 50-283-5721 Supplier MSC Supplier No. 48175970
This organic solvent degreaser is a plant-based, 100% petroleum-free, liquid concentrate with a citrus scent. It is a dilutable, water-based, and biodegradable formula designed for various cleaning applications.
  • Liquid form
  • Plastic drum container
  • 55 gallon container size
  • Citrus scent
  • Dilutable concentrate solution
  • Biodegradable, water-based formula
  • 40% VOC content
  • Distilled from all organic plant material
  • 100% free from petroleum solvents