Learn More
Medchemexpress LLC Oric-101 | 2222344-98-9 | 99.3% | 501.74 g·mol⁻1 | C34H47NO2 | 5 MG
SDP
Catalog No. 5000262252
Encompass_Preferred
ORIC-101 is a steroidal glucocorticoid receptor (GR) antagonist supplied as a high-purity solid for research use. It is intended for preclinical and cellular assays to study GR signaling and assess anti-cancer activity; molecular formula C34H47NO2, molecular weight 501.74 g·mol⁻1.
  • High purity appropriate for biochemical and cellular assays.
  • Solid, white to off-white powder for convenient handling.
  • Recommended storage maintains stability for long-term use.
  • Suitable for glucocorticoid receptor signaling and oncology research.
  • Documentation available: datasheet, COA, and SDS.
Catalog No. 50-002-62252 Supplier Medchemexpress LLC Supplier No. HY1127105MG
ORIC-101 is a steroidal glucocorticoid receptor (GR) antagonist supplied as a high-purity solid for research use. It is intended for preclinical and cellular assays to study GR signaling and assess anti-cancer activity; molecular formula C34H47NO2, molecular weight 501.74 g·mol⁻1.
  • High purity appropriate for biochemical and cellular assays.
  • Solid, white to off-white powder for convenient handling.
  • Recommended storage maintains stability for long-term use.
  • Suitable for glucocorticoid receptor signaling and oncology research.
  • Documentation available: datasheet, COA, and SDS.