Learn More
Medchemexpress LLC HY-N0004 10mg Medchemexpress, Oridonin CAS:28957-04-2 Purity:>98%
SDP
Catalog No. 501933347
Encompass_Preferred
Medchemexpress, HY-N0004 10mg Oridonin CAS:28957-04-2 Oridonin (NSC-250682), a diterpenoid isolated from Rabdosia rubescens, acts as an inhibitor of AKT , with IC 50 s of 8.4 and 8.9 μM for AKT1 and AKT2; Oridonin possesses anti-tumor, anti-bacterial and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3347 Supplier Medchemexpress LLC Supplier No. HYN000410MG
Medchemexpress, HY-N0004 10mg Oridonin CAS:28957-04-2 Oridonin (NSC-250682), a diterpenoid isolated from Rabdosia rubescens, acts as an inhibitor of AKT , with IC 50 s of 8.4 and 8.9 μM for AKT1 and AKT2; Oridonin possesses anti-tumor, anti-bacterial and anti-inflammatory effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.