Learn More
Medchemexpress LLC Oridonin | 28957-04-2 | 99.9% | 364.43 | 200 MG
SDP
Catalog No. 5000431626
Encompass_Preferred
Oridonin is a diterpenoid isolated from *Rabdosia rubescens*. It acts as an inhibitor of AKT, with IC50s of 8.4 μM for AKT1 and 8.9 μM for AKT2. Oridonin also exhibits anti-tumor, anti-bacterial, and anti-inflammatory effects. It is intended for research use only.
  • Inhibits AKT1 (IC50: 8.4 μM) and AKT2 (IC50: 8.9 μM)
  • Exhibits anti-tumor effects
  • Exhibits anti-bacterial effects
  • Exhibits anti-inflammatory effects
Catalog No. 50-004-31626 Supplier Medchemexpress LLC Supplier No. HYN0004200MG
Oridonin is a diterpenoid isolated from *Rabdosia rubescens*. It acts as an inhibitor of AKT, with IC50s of 8.4 μM for AKT1 and 8.9 μM for AKT2. Oridonin also exhibits anti-tumor, anti-bacterial, and anti-inflammatory effects. It is intended for research use only.
  • Inhibits AKT1 (IC50: 8.4 μM) and AKT2 (IC50: 8.9 μM)
  • Exhibits anti-tumor effects
  • Exhibits anti-bacterial effects
  • Exhibits anti-inflammatory effects