Learn More
Enzo Life Sciences Oridonin (50mg). CAS: 28957-04-2
SDP
Supplier:  Enzo Life Sciences ALX350390M050
Encompass_Preferred
Diterpene with potent anti-tumor activity. Induces apoptosis and autophagy. Alternative name: Isodonol. Purity: ≥98% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in DMSO (20mg/ml) or PEG 400; insoluble in water. Long Term Storage: +4°C.
Catalog No. 50-200-8877