Learn More
Selleck Chemical LLC Oridonin 10mg 28957-04-2 Isodonol, Rubescenin, NSC-250682
SDP
Supplier:  Selleck Chemical LLC S233510MG
Encompass_Preferred
Oridonin (Isodonol, Rubescenin, NSC-250682), a diterpenoid purified from Rabdosia rubescens, is a traditional agent with antitumor, anti-bacterial and anti-inflammatory effects. Oridonin inhibits AKT1 and AKT2 kinase activity with IC50 of 8.4 ?M and 8.9 ?M, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-667-8