Learn More
Tinius Olsen Testing Machine ORIFICE, CARBIDE (ASTM D1238)
SDP
Catalog No. NC9069799
Encompass
Orifice, MFI; Carbide, Standard 0.0825 in. ID x 0.315 in. Lg (ASTM D1238)
Catalog No. NC9069799 Supplier Tinius Olsen Testing Machine Supplier No. 02001031
Orifice, MFI; Carbide, Standard 0.0825 in. ID x 0.315 in. Lg (ASTM D1238)