Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Biology Bundle
SDPSCI_ED

Catalog No. S05724 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05724 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05724 Supplier EDVOTEK™ Supplier No. EVTBIOLOGY BUNDLE

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Biology Bundle contains all 83 of the biology Origami Organelles in one convenient set.

  • Teach Biology Using Origami. Download and print as many times as you want - forever at no extra cost.
  • Contains downloadable instructions and templates and PowerPoint presentations for the following Origami Organelles models:
    • Abdomen, Alveoli, Amoeba, Apoptosis, ATP Synthase, Anaerobic Respiration and Fermentation, Antibodies, Biochemistry of Vision, Calvin Cycle, Carbon Cycle, Cell Membranes, Cell Nucleus, Cell Signaling, Chlamydomonas, Citric Acid Cycle, Colorful Chromosomes, Cool Chloroplasts, CRISPR-Cas9, Crucial Krebs and the Link Reaction, Digestive System, DNA Replication, DNA Structure, Easy Eye, Electron Transport Chain, Endoplasmic Reticulum, Enzymes, Euglena, Fermentation, Fertilization and Reproductive Technology, Frog Life Cycle, Fun with Flowers, Gene Cloning, Gluconeogenesis, Glucose Uptake by Small Intestine, Glycolysis, Golgi and Lysosome, Groovy Glycolysis, Human Brain, Human Ear, Human Eye, Human Heart, Human Kidney, Immune System, Lac operon, Leaf Structure, Link Reaction/Pyruvate Oxidation, Liver and Spleen, Lungs, Lymph Node, Malaria, Mega Mitochondria, Mighty Mitochondria, Mitosis and Meiosis, Muscle Shuffle, Nephron, Nerve Cell, Nerve Impulses, Neuromuscular Junctions, Osmosis and Diffusion, Paramecium, Parasympatheic Nervous System, Phloem and Translocation, Photosynthesis Light Reactions, Polymerase Chain Reaction (PCR), Receptor Tyrosine Kinases (RTKs), Rod Photoreceptor Cell, Simple Animal Cell, Simple Bacterial Cell, Simple Plant Cell, Skin, Spinal Nerves, Stem Cells, Super Simple Cell, Sympathetic Nervous System, Synapses, Teeth, Transcription, Translation and Protein Synthesis, Urinary System, Villus, Water Cycle, Xylem, Zika Virus
  • Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
  • Download Includes:
    • Instructions and color and mono model templates.
    • PowerPoint classroom presentations are included for all models.
    • Due to the number of files in this set, 9 email links are sent within a few hours of purchase.
  • All You Need: Paper, scissors, glue or tape.

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.