Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Climate Change Model
SDPSCI_ED

Catalog No. S05718 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05718 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05718 Supplier EDVOTEK™ Supplier No. EVT616

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Learn about climate change in an engaging way with the easy to make Origami Organelles Climate Change Model.

  • Make the model using the colorful parts.
  • Use the model to look at how the climate has changed due to human activities.
  • The model is a starting point to consider how climate change will effect us and what we can do to help solve the environmental problems it has caused and will cause in the future.
  • This unique model covers: rising sea levels, sea ice melting, glacial retreat, wildfires, pollution, extreme weather events, habitat loss, deforestation, desertification, ocean acidification, disruption to ecosystems and threats to low level coastlines.
  • Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
  • Dimensions: 23 cm x 25 cm x 10 cm (9 in. x 10 in. x 4 in.)
  • Download includes: instructions, color and mono model templates, and Powerpoint presentation.
  • All you need: paper, scissors and tape.

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Earth Science - Weather and Climate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.