Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles CRISPR Model
SDPSCI_ED

Catalog No. S05734 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05734 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05734 Supplier EDVOTEK™ Supplier No. EVT031

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Give your students an insight into genome editing with the Origami Organelles CRISPR Model.

  • CRISPR is a revolutionary new genetic engineering technique that makes editing genomes easy and inexpensive.
  • It is based on a type of immune system found in many types of prokaryote.
  • Students first make 3D models of the components of CRISPR - the enzyme Cas9 and guide RNAs.
  • Next, they use these to see how genome editing is done. Making the model and carrying out the activity makes CRISPR easy to understand.
  • Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
  • Dimensions: 26 cm x 18 cm (10 in. x 7 in.)
  • Download includes: instructions, model templates, and full color PowerPoint class presentation
  • All you need: paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.