Learn More
Description
- With this engaging hands-on set, students use easy-to-make models of the main enzymes involved in DNA replication (DNA polymerase, helicase and primase) to copy a DNA template.
- They learn about the importance of RNA primers, leading and lagging strands, and can even do an optional activity on proofreading.
- Resource for advanced or introductory level lessons.
- Takes about 20 minutes to make a 12 base pair model.
- Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.
- Dimensions: 35 cm (14 in.) long
- Download includes: instructions, model templates, and PowerPoint file.
- All you need: paper, scissors, and tape.
Specifications
Specifications
| Grade | 4 to 12 |
| Format | Model |
| Languages | English |
| Subject | Biology - Life Science |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

