Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Earth Science Bundle
SDPSCI_ED

Catalog No. S05714 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
75 Students
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S05714 75 Students
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05714 Supplier EDVOTEK™ Supplier No. EVT218

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This set contains 27 of the Earth Science Origami Organelles in one convenient bundle. Contains downloadable instructions, templates, and classroom PowerPoint presentations for these 27 Origami Organelles models.

  • Teach Earth Science with downloadable paper models.
  • Using the unique Origami Organelles sets, your students will see Earth Science in a new way.
  • This set contains 27 of our Earth Science Origami Organelles in one convenient bundle.
  • Contains downloadable instructions, templates, and classroom PowerPoint presentations for these 27 Origami Organelles models:
    • Braided Streams
    • Carbon Cycle
    • Climate Change
    • Death Valley
    • Desert Formation
    • Desert Landforms
    • Drainage Basins
    • Earthquakes
    • Estuaries
    • Floodplains
    • Folding Rocks
    • Glaciers
    • Grand Canyon
    • Igneous Rocks
    • Interlocking Spurs
    • Life Cycle of a Star
    • Meanders
    • Minerals
    • Oxbow Lakes
    • River Valleys
    • Rock Cycle
    • Structure of the Earth
    • Structure of the Sun
    • Tectonic Plates
    • Volcanoes
    • Water Cycle
    • Waterfalls and Gorges
  • Each model takes about 30 minutes to make.
  • Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Specifications

Grade 4 to 12
Includes 27 Earth Science Origami Organelles
Class Size 75 Students
Product Type Origami Organelles Earth Science Bundle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.