Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Gene Cloning Model
SDPSCI_ED

Catalog No. S05727 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05727 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05727 Supplier EDVOTEK™ Supplier No. EVT078
This item is not returnable. View return policy

Gene cloning is easy to grasp with the colorful Origami Organelles Gene Cloning Model.

  • With this interactive model your students create base pairs and combine them to make a model of plasmid.
  • Learn about gene cloning or genetic engineering by carrying out a restriction enzyme digest and adding a gene insert.
  • nsert can represent either the insulin or green fluorescent protein (GFP) genes.
  • Students learn about plasmid structure, use of vectors, marker genes, restriction enzymes recognition sites, sticky ends, multiple cloning sites, and promoters.
  • Takes about 20 minutes to make if divided between 5 groups of students. Can be kept intact and reused once assembled.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
40 cm (16 in.) diameter

Download includes:
Instructions, color and mono model templates and PowerPoint file

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.