Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Lac Operon Model
SDPSCI_ED

Catalog No. S05728 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05728 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05728 Supplier EDVOTEK™ Supplier No. EVT071

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Gene regulation in E. coli made clear with the Origami Organelles Lac Operon Model.

  • The lac operon consists of 3 genes that the bacteria E. coli use to utilize lactose as a food source. The genes are only switched on when lactose is present.
  • Your students will make a model of the lac operon and use it to see how bacteria are able to control these genes. The model covers the lac repressor, RNA polymerase, operator, promoter and the lacZ, lacY and lacA genes and their products.
  • Makes an engaging way to explain prokaryotic gene regulation.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
48 cm x 18 cm (19 in. x 7 in.)

Download includes:
Instructions, model templates, and PowerPoint file

All you need:
Scissors and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.