Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Mitosis and Meiosis Model
SDPSCI_ED

Catalog No. S05731 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05731 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05731 Supplier EDVOTEK™ Supplier No. EVT036

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Origami Organelles Mitosis and Meiosis Model offers a way to teach mitosis and meiosis.

  • Students make colorful model chromosomes and use them to work through the stages of cell division. The model can be used to teach mitosis and meiosis.
  • Students cover the stages of mitosis, including chromosome alignment and segregation. They will also learn about the cell cycle as they make two diploid daughter cells.
  • Students can then carry out the stages of meiosis, including independent assortment and crossing over, whilst creating four haploid daughter cells.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
Largest chromosome 12 cm (5 in.) long

Download includes:
Instructions, model templates, and PowerPoint file

All you need:
Scissors and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.