Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Nitrogen Cycle Model
SDPSCI_ED

Catalog No. S05717 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05717 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05717 Supplier EDVOTEK™ Supplier No. EVT901
This item is not returnable. View return policy

Show your class the nitrogen cycle with the Origami Organelles Nitrogen Cycle Model.

Your students will enjoy learning about the nitrogen cycle with our interactive model.

  • Students build their model using the colorful parts of the template
  • Discover the features of the nitrogen cycle by visualizing connections as they make the model
  • Takes about 30 minutes to build the model.

The model covers: nitrogen fixation (legumes, lightning and Haber-Bosch process), assimilation, ammonification (decay), nitrification, denitrification and combustion of fossil fuels. It also considers the human impact on the nitrogen cycle.

It can even be made into a colorful 3-dimensional classroom poster.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
67 cm x 50 cm (26 in. x 20 in.)

Download includes:
Instructions, templates, and classroom PowerPoint presentation

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.