Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Plant Life Cycle Model
SDPSCI_ED

Catalog No. S05716 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05716 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05716 Supplier EDVOTEK™ Supplier No. EVT331
This item is not returnable. View return policy

Show how plant life cycles work with the Origami Organelles Plant Life Cycle Model.

The life cycle of a flowering plant takes it from a seed to a fully grown plant and back to a seed again. This model shows the stages of a plant life cycle and what happens at each step.

  • First, students build their models of a seed, seedling, tree, flower, bee, fruit and deer.
  • Utilize the models to learn about the germination, growth, flowering, pollination, fertilization, fruit formation, and seed dispersal
  • Takes just 30 minutes to build the model.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions (flower):
12 cm x 10 cm (5 in. x 4 in.)

Download includes:
Instructions, color and mono model templates, and classroom PowerPoint file

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Plants
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.