Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Polymerase Chain Reaction (PCR) Model
SDPSCI_ED

Catalog No. S05735 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05735 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05735 Supplier EDVOTEK™ Supplier No. EVT032
This item is not returnable. View return policy

Teach PCR with the Origami Organelles Polymerase Chain Reaction (PCR) Model.

  • Help your students to understand PCR with this colorful model. Students make models of Taq DNA polymerase and use it to extend primers on template DNA. They can carry out as many cycles as they like with the components provided.
  • They learn about the importance of Taq polymerase, that DNA is only synthesized in the 5' to 3' direction, DNA primers, the steps of PCR - denaturing, annealing and extension. Compare PCR and DNA replication using the DNA replication model.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
DNA Template 35 cm (14 in.) long

Download includes:
Instructions, model templates, and PowerPoint file.

All you need:
Paper, scissors, and tape.

Specifications

Grade 6 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.