Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Simple Bacterial Cell Model
SDPSCI_ED

Catalog No. S05722 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05722 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05722 Supplier EDVOTEK™ Supplier No. EVT035
This item is not returnable. View return policy

Build a beautiful bacterial cell model with the Origami Organelles Simple Bacterial Cell Model.

  • Bacterial cells are one of the oldest forms of life on Earth and cause many human diseases. Give your students an insight into the structure of these important organisms.
  • The model shows your students the main structures of a bacterial cell: cell wall, cell membrane, genome, plasmid, flagellum. This easy to make model does not need any tape or glue to build.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
17 cm x 5 cm (7 in. x 2 in.

Download includes:
Instructions, model templates, and full color PowerPoint class presentation

All you need:
Paper and scissors

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Cells
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.