Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Synapses Model
SDPSCI_ED

Catalog No. S05740 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
Catalog No. Grade
S05740 4 to 12
1 options
This item is not returnable. View return policy

Catalog No. S05740

Supplier: EDVOTEK™ EVT006

This item is not returnable. View return policy

Use the Origami Organelles Synapses Model to show your students synapses.

  • Synapses become easy to understand with this membrane modeling system.
  • After making their models, your students will fuse a synaptic vesicle to the presynaptic membrane and release a neurotransmitter. Once this crosses the synaptic cleft and binds to the receptor, ion channels open and cause changes in the postsynaptic membrane that may lead to an action potential if the threshold is passed.
  • Covers synapse structure, role of neurotransmitter, summation, inhibitory and excitatory synapses.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
25 cm (10 in.) across

Download includes:
Instructions, color and mono model templates, and PowerPoint file.

All you need:
Paper, scissors, and tape.

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.