Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Origami Organelles Transcription Model
SDPSCI_ED

Catalog No. S05729 Shop All EDVOTEK Products
Change view
Click to view available options
Grade:
4 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S05729 4 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S05729 Supplier EDVOTEK™ Supplier No. EVT070
This item is not returnable. View return policy

Use the Origami Organelles Transcription Model to teach transcription.

  • Transcription is how the genetic information stored as DNA in the nucleus is copied into RNA so it can be used to make proteins.
  • Your students will make a model of a DNA template and transcribe it into messenger RNA. The model covers the role of promoters, transcription factors, and RNA polymerase. RNA processing is also covered including RNA splicing, exons, introns, poly-A-tail and 5' cap.
  • This colorful model shows transcription in a clear way.

Origami Organelles are downloadable paper models that you print and make as many times as you like. When you purchase a model, provide an email address for the PDF to be emailed to and then you are licensed for unlimited use on a single site or campus.

Dimensions:
64 cm x 26 cm (25 in. x 10 in.)

Download includes:
Instructions, model templates, and PowerPoint file

All you need:
Paper, scissors, and tape

Specifications

Grade 4 to 12
Format Model
Languages English
Subject Biology - Life Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.