Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OriGene OriGene qPCR Primer Pair, CCR6 Human, NM_004367, 200 Reactions
SDP

Catalog No. NC3992919
Encompass_Preferred

This qPCR primer pair is designed for the detection of the Homo sapiens CCR6 gene using qSTAR technology. It comes as a lyophilized primer mix sufficient for 200 reactions, offering reliability and convenience for various molecular biology applications.

  • qSTAR qPCR primer pairs against Homo sapiens gene CCR6.
  • Sufficient for 200 reactions.
  • Primer mix is lyophilized.
  • Reconstitute in 200 μL dH2O to make a final concentration of 10 μM.
  • Quality controlled to generate satisfactory qPCR data on ABI 7900HT.
  • Store at -20°C.
  • Stable for one year from date of shipping.

Catalog No. NC3992919 Supplier OriGene Supplier No. HP207690
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This qPCR primer pair is designed for the detection of the Homo sapiens CCR6 gene using qSTAR technology. It comes as a lyophilized primer mix sufficient for 200 reactions, offering reliability and convenience for various molecular biology applications.

  • qSTAR qPCR primer pairs against Homo sapiens gene CCR6.
  • Sufficient for 200 reactions.
  • Primer mix is lyophilized.
  • Reconstitute in 200 μL dH2O to make a final concentration of 10 μM.
  • Quality controlled to generate satisfactory qPCR data on ABI 7900HT.
  • Store at -20°C.
  • Stable for one year from date of shipping.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.