Learn More
OriGene OriGene qPCR Primer Pair, Human CCR4, NM_005508, 200 Reactions
SDP
Catalog No. NC3992918
Encompass_Preferred
This qSTAR qPCR primer pair is designed to target the Homo sapiens gene CCR4. It comes as a lyophilized primer mix, sufficient for 200 reactions, enabling quantitative PCR for human CCR4 gene expression analysis. The primer mix has been quality controlled and tested to generate satisfactory qPCR data on ABI 7900HT, and is stable for one year from the date of shipping when stored at -20°C.
  • qSTAR qPCR primer pairs
  • Targets Homo sapiens gene CCR4
  • Lyophilized primer mix
  • Sufficient for 200 reactions
  • Quality controlled for satisfactory qPCR data on ABI 7900HT
  • Stable for one year from date of shipping when stored at -20°C
  • Enables quantitative PCR for human CCR4 gene expression analysis
  • Tested and validated for performance on ABI 7900HT
Catalog No. NC3992918 Supplier OriGene Supplier No. HP208627
This qSTAR qPCR primer pair is designed to target the Homo sapiens gene CCR4. It comes as a lyophilized primer mix, sufficient for 200 reactions, enabling quantitative PCR for human CCR4 gene expression analysis. The primer mix has been quality controlled and tested to generate satisfactory qPCR data on ABI 7900HT, and is stable for one year from the date of shipping when stored at -20°C.
  • qSTAR qPCR primer pairs
  • Targets Homo sapiens gene CCR4
  • Lyophilized primer mix
  • Sufficient for 200 reactions
  • Quality controlled for satisfactory qPCR data on ABI 7900HT
  • Stable for one year from date of shipping when stored at -20°C
  • Enables quantitative PCR for human CCR4 gene expression analysis
  • Tested and validated for performance on ABI 7900HT