Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Murphy Oil Soap Wood Cleaner, Original, 32 oz Bottle, 9/Carton

Catalog No. 502817372
Encompass

Murphy Oil Soap Wood Cleaner, Original, is specifically formulated to clean finished wood and non-wood surfaces. This multi-purpose cleaner helps maintain a natural shine and enhances the beauty of wood in a gentle way, utilizing 98% natural ingredients.

  • Contains 98% natural ingredients
  • Free from ammonia, bleach, and phosphates
  • Biodegradable formulation
  • Safely cleans wood surfaces
  • Delivers a natural shine
  • Trusted for over 100 years
  • PH of 11.0
  • Contains 1–5% sodium hydroxide and 0.1–1% citronella oil

Catalog No. 50-281-7372 Supplier MSC Supplier No. 07307424
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Murphy Oil Soap Wood Cleaner, Original, is specifically formulated to clean finished wood and non-wood surfaces. This multi-purpose cleaner helps maintain a natural shine and enhances the beauty of wood in a gentle way, utilizing 98% natural ingredients.

  • Contains 98% natural ingredients
  • Free from ammonia, bleach, and phosphates
  • Biodegradable formulation
  • Safely cleans wood surfaces
  • Delivers a natural shine
  • Trusted for over 100 years
  • PH of 11.0
  • Contains 1–5% sodium hydroxide and 0.1–1% citronella oil

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.