Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drummond™ Original Pipet-Aid™ Pipet Controller

Catalog No. 1368115C
Change view
Click to view available options
Model:
Complete (Dual Pump, TC Nosepiece), CE/Eur
Standard, CE/Eur
TC Nosepiece
Voltage:
110 V
220 V
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Voltage
13-681-15C TC Nosepiece 110 V
13-681-15A Standard, CE/Eur 220 V
13-681-10 Complete (Dual Pump, TC Nosepiece), CE/Eur 220 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13-681-15C Supplier Drummond™ Supplier No. 4000110TC

missing translation for 'validMainframeMsg'

Sets the standard for a safe and convenient alternative to mouth pipetting in the laboratory.

A patented, easy-to-use volume control mechanism permits precise fluid intake and delivery. The check valve protects against accidental fluid intake into the handle.

  • Can be used with glass or plastic pipets
  • Autoclavable components
  • Optional EZ Grip Tissue Culture (TC) Nosepiece prevents cross-contamination due to over-pipetting
  • Special dual pump filtration unit for use with biohazardous materials is available
  • Standard 110V models for use in the U.S. and 220V CE models for use in Europe

† - Complete models include Pipet-Aid, TC nosepiece, four extra filters and dual-pump filtration unit

Order Info

Complete models include Pipet-Aid, TC nosepiece, four extra filters and dual-pump filtration unit. Contact your Fisher Scientific Customer Service Representative for available replacement parts.

Specifications

Autoclavable Autoclavable
Model TC Nosepiece
Type Filler/Dispenser
Volume (Metric) 1 to 150 mL
Filter Type Standard
For Use With (Application) Pipetting of biohazardous materials
For Use With (Equipment) Glass or plastic pipets
Voltage 110 V
Frequency 60 Hz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.