Learn More
Grainger O-Ring, 014, 713P71
Catalog No. NC2880650
Encompass_Preferred
The O-Ring 014-713P71 is a 70 durometer nitrile rubber O-ring with an inside diameter of 0.139 inches, an outside diameter of 0.187 inches, and a width of 0.024 inches. It has a temperature range of -30°F to +200°F and is rated for continuous use with petroleum oils. This O-ring meets military specification MIL-P-25732 and is made in the USA by Marco Rubber. It can provide a tight seal between machine parts and is sold in a package of 10 rings.
Catalog No. NC2880650 Supplier Grainger Supplier No. 713P71
The O-Ring 014-713P71 is a 70 durometer nitrile rubber O-ring with an inside diameter of 0.139 inches, an outside diameter of 0.187 inches, and a width of 0.024 inches. It has a temperature range of -30°F to +200°F and is rated for continuous use with petroleum oils. This O-ring meets military specification MIL-P-25732 and is made in the USA by Marco Rubber. It can provide a tight seal between machine parts and is sold in a package of 10 rings.