Learn More
Perkin Elmer US LLC Internal Injector Support Adapter O-Ring, 14.0 mm I.D., for ELAN/NexION 300/350
Catalog No. 509052273
Encompass_Preferred
This O-ring is included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100) and the Injector Adapter Kit for NexION 300/350 (N8140527).
Catalog No. 50-905-2273 Supplier Perkin Elmer US LLC Supplier No. N8122010
This O-ring is included in the ELAN Cassette O-Ring Kit for Injector Support for ELAN (N8120100) and the Injector Adapter Kit for NexION 300/350 (N8140527).