Learn More
Perkin Elmer US LLC O-Ring Free Glass Baffled Cyclonic Spray Chamber for Avio 200/500
Catalog No. 502113602
Encompass_Preferred
This baffled cyclonic spray chamber, made of borosilicate glass, is an integral part of the sample introduction system responsible for filtering the sample mist to permit appropriate droplet size distribution to reach the plasma. The spray chamber features a sand-blasted neck with EZ seal fitting and includes a drain fitting and metal clip.
Catalog No. 50-211-3602 Supplier Perkin Elmer US LLC Supplier No. N0791352
This baffled cyclonic spray chamber, made of borosilicate glass, is an integral part of the sample introduction system responsible for filtering the sample mist to permit appropriate droplet size distribution to reach the plasma. The spray chamber features a sand-blasted neck with EZ seal fitting and includes a drain fitting and metal clip.