Learn More
Perkin Elmer US LLC O-Ring Free Quartz Baffled Cyclonic Spray Chamber for NexION 300/350
Catalog No. 502104807
Encompass_Preferred
This baffled cyclonic spray chamber, made of quartz, is designed for all-purpose, non-hydrofluoric acid applications. It provides excellent sensitivity, stability and low blanks.
Catalog No. 50-210-4807 Supplier Perkin Elmer US LLC Supplier No. N8145013
This baffled cyclonic spray chamber, made of quartz, is designed for all-purpose, non-hydrofluoric acid applications. It provides excellent sensitivity, stability and low blanks.