Learn More
Chemglass Life Sciences Silicone O-Ring for 9mm Compression Fitting, FDA Grade
SDP
Catalog No. 50143775
Encompass
Orange silicone o-rings conform to FDA 21CRF Part 177.2600.
Catalog No. 50-143-775 Supplier Chemglass Life Sciences Supplier No. CLS1705110
Orange silicone o-rings conform to FDA 21CRF Part 177.2600.