Learn More
Thermo Scientific™ Orion™ AQUAfast 24mm Round Vials for Colorimeters and Spectrophotometers
Shop All Thermo Scientific ProductsDescription
Use Thermo Scientific™ Orion™ AQUAfast 24mm Round Vials with a variety of Orion™ colorimeters and spectrophotometers. The Orion™ AQUAfast 24mm Round Vials can be used with Orion™ AQUAfast colorimetric powder pack, tablet and liquid reagents to measure a variety of common water and wastewater parameters. Catalog number AC2V24 contains 12 x 24mm vials.
Specifications
Specifications
| Description | Round vials |
| Diameter (Metric) Outer | 24 mm |
| Quantity | 12 Cs. |
| Certifications/Compliance | Certificate of Analysis (COA) |
| Product Type | Vial |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.