Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Orion™ 8102BNUWP ROSS Ultra™ pH Electrode PROMO

Catalog No. 13642237PMO Shop All Thermo Scientific Products
Change view
Click to view available options
Connector Types:
BNC (Waterproof)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connector Types
13-642-237PMO BNC (Waterproof)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13-642-237PMO Supplier Thermo Scientific™ Supplier No. 8102BNUWPPM

When you purchase a Thermo Scientific™ ROSS™ pH Electrode, you’ll receive a ROSS Buffer Kit at no additional cost. Order using the catalog numbers below and your products will ship together.

Includes:

  • 60 mL bottle of Orion ROSS electrode fill solution, 810007

Requires:

  • pH buffers for calibration
  • Beakers, plastic or glass
  • Magnetic stirrer or Orion stirrer probe
  • Distilled or deionized water

Compatible with:

Waterproof BNC connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.

Recommended for:

  • Q.C. and research applications
TRUSTED_SUSTAINABILITY

Specifications

Description Glass body combination electrode
Connector Types BNC (Waterproof)
Junction Ceramic
Material Glass Body
pH Range 0 to 14
Temperature (Metric) Operating 0°C to 100°C
Type pH Electrode
Drift <0.002 pH/day
Filling Solution Type 3M KCl
Includes 60 mL bottle of Orion ROSS electrode fill solution
Internal Reference ROSS
Precision 0.01
Temperature (English) Operating 32 to 212°F
Length (English) Cable 39.4 in.
Length (Metric) Cable 1 m
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.