Learn More
Thermo Scientific™ Orion™ ROSS Ultra™ Thin Stem Glass Bodied Combination pH Electrode
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Orion™ ROSS Ultra™ Glass Semi-Micro Electrodes provide optimum accuracy and unmatched response in samples of varying temperatures. They are available in chemically resistant glass bodies and are best suited for measuring samples in test tube and small containers. Orion™ ROSS Ultra™ Glass Semi-Micro Electrodes come with a two year warranty.
Orion™ ROSS Ultra™ Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
Solutions required
- Electrode fill solution
- 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles
Electrode storage solution
- 810001 ROSS pH electrode storage solution, 475 mL bottle
pH buffers
- 910168 pH 1.68 buffer, NIST traceable, 475 mL
- 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
- 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
- 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
- 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
- 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
- 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
- 910112 pH 12.46 buffer, NIST traceable, 475 mL
- 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle
pH cleaning solution kit
- 900020 Cleaning solution A and C, 30 mL each. B and D, 60 ml each
Items included with
- 810007 ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
- 24 months from date of purchase
Specifications
Specifications
| Description | Semi-micro tipped glass combination electrode; 165 x 6mm |
| Connector Types | BNC (Waterproof) |
| Junction | Ceramic |
| Material | Glass Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | pH Electrode |
| Filling Solution Type | 3M KCl |
| For Use With (Equipment) | Standard pH Meter |
| Includes | 60ml bottle of Orion ROSS electrode fill solution |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.