Learn More
Thermo Scientific™ Orion™ 8135BNUWP ROSS Ultra™ pH Electrode PROMO
Description
Includes:
- 1 m cable
Requires:
- pH buffers (pH 7 ±1 to 3 pH units), beakers, plastic or glass
- Magnetic stirrer or Orion stirrer probe
- Distilled or deionized water
Compatible with:
BNWP connector is compatible with any standard BNC connector both on Thermo Scientific™ Orion Star™ Series meters and competitive meter models.
Recommended for:
- Measuring pH of soft, moist surfaces both solid and semi-solid such as agar gel plates, meats, bread dough and similar samples
Specifications
Specifications
| Description | Ultra pH Electrode |
| Connector Types | BNC |
| Junction | Glass Fiber |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | pH Electrode |
| Electrode | Flat surface tip epoxy body combination electrode; 20 x 12 mm |
| Filling Solution Type | 3M KCl |
| Internal Reference | ROSS |
| Show More |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.