Learn More
Thermo Scientific™ Orion™ Sure-Flow™ Single Junction Epoxy Bodied Reference Electrode
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Orion™ Single Junction Electrodes provide a reliable option for routine pH measurement. These silver chloride electrodes are available in a variety of styles and provide accurate measurements of typical samples.
Solutions required
- Electrode fill solution
- 900001 Equitransferent solution with Ag/AgCl, 5 x 60 mL
- For dilute samples < 0.2 M in total ionic strength and temperatures > 10 °C.
- 900011 4 M KCl with Ag/AgCl, 5 x 60 mL
- For concentrated samples and temperatures < 10 °C.
- For complete details regarding the proper choice of reference electrode filling solution, refer to the user guide for the ion selective electrode or pH electrode to be used.
pH buffers
- 910168 pH 1.68 buffer, NIST traceable, 475 mL
- 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
- 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
- 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
- 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
- 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
- 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
- 910112 pH 12.46 buffer, NIST traceable, 475 mL
- 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle
pH cleaning solution kit
- 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each
Items included with
- 810007 ROSS pH electrode fill solution 60 mL 1 bottle
Warranty period
- 12 months from date of purchase
Specifications
Specifications
| Description | 900100 Sure-Flow Reference Half Cell Electrode |
| Connector Types | BNC |
| Junction | Sure-Flow Sleeve Junction |
| Material | Epoxy |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 100°C |
| Type | Half Cell Reference Electrode |
| Electrode | Single Junction Ref Electrode |
| Filling Solution Type | Orion 900001 Filling Solution |
| For Use With (Application) | ISE determinations |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.