Learn More
Thermo Scientific™ Orion™ Double Junction Epoxy Bodied Combination pH Electrode
Shop All Thermo Scientific ProductsDescription
Orion™ Double Junction Electrodes have an isolated Ag/AgCl reference system which prevents silver from coming in contact with the sample. Great for measuring TRIS buffer, sulfide and protein samples.
Solutions required
- Electrode fill solution
- 910008 3M KCl filling solution, 5 x 60 mL bottles
Electrode storage solution
- 910001 pH electrode storage solution, 475 mL bottle
pH buffers
- 910168 pH 1.68 buffer, NIST traceable, 475 mL
- 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
- 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
- 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
- 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
- 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
- 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
- 910112 pH 12.46 buffer, NIST traceable, 475 mL
- 910199 Standard All-in-One pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each, pH
- electrode storage solution, 475 mL, and 12 mm electrode storage bottle
pH cleaning solution kit
- 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each
Items included with
- 910008 3M KCl filling solution 60 mL 1 bottle
Warranty period
- 12 months from date of purchase
Specifications
Specifications
| Description | Double Junction pH Electrode |
| Connector Types | BNC (Waterproof) |
| Junction | Wick |
| Material | Epoxy Body |
| pH Range | 0 to 14 |
| Temperature (Metric) Operating | 0°C to 60°C |
| Type | Double Junction pH Electrode |
| For Use With (Equipment) | Standard pH Meter |
| Internal Reference | Ag/AgCl Double Junction |
| Precision | 0.02 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.