Learn More
Description
- All Orion pH Buffer Bottles have pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- pH 4.01 buffer is pink
- pH 5.00 buffer is orange
- pH 7.00 buffer is yellow
- pH 10.01 buffer is blue
- Buffers are available in 475 mL (1 pint) bottles, portable five packs of 5 x 60 mL (2 oz.) bottles, convenient All-in-One buffer kits, 19 L (5 gal.) Cubitainers™ and single-use pouches
Specifications
Specifications
| For Use With (Application) | Calibration |
| pH | 4.01, 7, 10.01 |
| Quantity | 5 x 60 mL |
| Packaging | Kit |
| Traceability to NIST | Yes |
| Includes | 60 mL Bottle of pH 4.01, 7 and 10.01 Buffers, 60 mL Bottle of pH Electrode Storage Solution, and 60 mL Bottle of pH Electrode Cleaning Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.