Learn More
Thermo Scientific™ Orion™ AQ4500 Turbidimeter
Catalog No. 13650100 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Turbidity Meter
Catalog No. Type
13-650-100 Turbidity Meter
1 options
Reorder replacement Calibration Standards for AQ4500 (AC45ST) and replacement vials in a package of 12 (AC2T24). Silicone oil and cloth set (AC45S1) is also available.
Catalog No. 13-650-100 Supplier Thermo Scientific™ Supplier No. AQ4500
  • $3,480.00
    $2,753.65 / Each of 1
    Save $726.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Reorder replacement Calibration Standards for AQ4500 (AC45ST) and replacement vials in a package of 12 (AC2T24). Silicone oil and cloth set (AC45S1) is also available.
Utilize dual LED light sources and drinking water/wastewater methods approved for U.S. EPA regulatory reporting offered by this turbidity meter.
Utilize advanced features with the Thermo Scientific™ Orion™ AQ4500 Turbidity Meter, including dual light sources and drinking water/wastewater methods approved for U.S. EPA regulatory reporting. Keep pace with changing regulatory protocols and use White Light LED to comply with EPA 180.1 requirements or Infrared (IR) LED to comply with ISO 7027 requirements. Select from measure modes including EPA 180.1, ISO Nephelometric, ISO Absorbance, % transmittance, EBC and ASBC. This IP67 waterproof meter records 100 data points and offers an average battery life of 2500 hours intermittent use.
Log data into internal memory, or order AQ4CBL cable to send data to printer or download to a computer for easy time- and date-stamped results storage.
  • U.S. EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg. 38351-38356
  • Dual source to comply with EPA 180.1 and ISO 7027
  • Nephelometric and ratiometric measurements with autoranging
  • IP67 waterproof portable turbidity meter
  • Readings in the range of 0 to 4000 NTU with a choice of units: NT, FTU, FNU, ASBC or EBC
Includes:
  • AQ4500 meter
  • Set of 5 USEPA approved primary calibration standards
  • Seven glass vials
  • User guide
  • Carrying case
TRUSTED_SUSTAINABILITY
Specifications
Type Turbidity Meter
Light Source LED, Multiple
Display Type LED
Automatic Range Selection Yes
Description Turbidity Meter and Calibration Kit
Battery Life 2500 Hours
EPA Approved Methods 180.1
Certifications/Compliance US EPA approval for Drinking Water Reporting per 40 CFR Part 141, vol. 74, no. 147, Monday, August 3, 2009. Rules and regulations pg 38351-38356.EPA 180.1 and ISO 7032
ISO Accepted Methods ISO 7027